5-(Trifluoromethyl)-4H-1,2,4-triazol-3-amine 95%

Supplier: Ambeed
Warning

Synonyms: 5-(Trifluoromethyl)-1H-1,2,4-triazol-3-amine

A281166-100MG A281166-10G A281166-250MG A281166-5G A281166-1G
77002-174EA 14.86 USD
77002-174 77002-176 77002-178 77002-180 77568-570
5-(Trifluoromethyl)-4H-1,2,4-triazol-3-amine 95%
5-(Trifluoromethyl)-4H-1,2,4-triazol-3-amine

Precaución: For research use only.

Formula: C₃H₃F₃N₄
MW: 152.08 g/mol
MDL Number: MFCD00524733
CAS Number: 25979-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR