Fmoc-L-4-methylphe 97%

Supplier: Ambeed
Warning

A280810-1G A280810-250MG A280810-25G A280810-5G A280810-10G A280810-100G
77002-162EA 23 USD
77002-162 77002-164 77002-166 77550-724 77601-238 77672-450
Fmoc-L-4-methylphe 97%
Fmoc-L-4-methylphe

Precaución: for Research Use Only

Formula: C₂₅H₂₃NO₄
MDL Number: MFCD00270197
CAS Number: 199006-54-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR