4-Formyl-3-nitrobenzonitrile 95%

Supplier: Ambeed
Warning

A280626-100MG A280626-1G A280626-250MG
77032-160EA 25 USD
77032-160 77032-162 77032-164
4-Formyl-3-nitrobenzonitrile 95%
4-Formyl-3-nitrobenzonitrile

Precaución: for Research Use Only

Formula: C₈H₄N₂O₃
MDL Number: MFCD10696869
CAS Number: 90178-78-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR