Sarcosine methyl ester hydrochloride 97%

Supplier: Ambeed
Warning

A280604-100G A280604-10G A280604-25G A280604-5G A280604-500G
77002-148EA 113 USD
77002-148 77002-150 77002-152 77002-154 77672-446
Sarcosine methyl ester hydrochloride 97%
Sarcosine methyl ester hydrochloride

Precaución: for Research Use Only

Formula: C₄H₉NO₂·HCl
MDL Number: MFCD00038876
CAS Number: 13515-93-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR