tert-Butyl-4-ethynylbenzoate 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 4-ethynylbenzoate

A280588-100MG A280588-250MG A280588-1G A280588-10G
77014-606EA 15 USD
77014-606 77014-608 77571-744 77890-510
tert-Butyl-4-ethynylbenzoate 97%
tert-Butyl-4-ethynylbenzoate

Precaución: for Research Use Only

Formula: C₁₃H₁₄O₂
MDL Number: MFCD09037869
CAS Number: 111291-97-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR