2-Bromo-3,6-difluorobenzaldehyde 98%

Supplier: Ambeed
Warning

A280200-100MG A280200-1G A280200-250MG A280200-5G A280200-25G
77014-596EA 6.37 USD
77014-596 77014-598 77014-600 77014-602 77683-845
2-Bromo-3,6-difluorobenzaldehyde 98%
2-Bromo-3,6-difluorobenzaldehyde

Precaución: For research use only.

Formula: C₇H₃BrF₂O
MW: 221.00 g/mol
MDL Number: MFCD12922618
CAS Number: 934987-26-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR