Fmoc-N-(2-boc-aminoethyl)-Gly-OH 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-N-(N-β-Boc-aminoethyl)-Gly-OH, Fmoc-N-(2-Boc-aminoethyl)-Gly-OH

A279949-100MG A279949-250MG
76988-466EA 53.05 USD
76988-466 76988-468
Fmoc-N-(2-boc-aminoethyl)-Gly-OH 97%
Fmoc-N-(2-boc-aminoethyl)-Gly-OH

Precaución: for Research Use Only

Formula: C₂₄H₂₈N₂O₆
MW: 440.496 g/mol
MDL Number: MFCD02259490
CAS Number: 141743-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR