tert-Butyl 3-(2-(tert-butoxy)-2-oxoethyl)-4-oxopiperidine-1-carboxylate 97%

Supplier: Ambeed
Warning

A279945-100MG A279945-1G A279945-250MG
77032-148EA 62 USD
77032-148 77032-150 77032-152
tert-Butyl 3-(2-(tert-butoxy)-2-oxoethyl)-4-oxopiperidine-1-carboxylate 97%
tert-Butyl 3-(2-(tert-butoxy)-2-oxoethyl)-4-oxopiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₆H₂₇NO₅
MDL Number: MFCD11111810
CAS Number: 1010814-94-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR