4-Amino-N-(tert-butoxycarbonyl)-D-phenylalanine 98%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-N-Boc-D-phenylalanine, Boc-D-Phe(4-NH₂)-OH, Boc-D-Phe(4-NH2)-OH

A279854-10G A279854-1G A279854-250MG A279854-5G A279854-25G
77002-126EA 53 USD
77002-126 77002-128 77002-130 77002-132 77601-234
4-Amino-N-(tert-butoxycarbonyl)-D-phenylalanine 98%
4-Amino-N-(tert-butoxycarbonyl)-D-phenylalanine

Precaución: for Research Use Only

Formula: C₁₄H₂₀N₂O₄
MW: 280.32 g/mol
MDL Number: MFCD00236818
CAS Number: 164332-89-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR