(4-Bromophenyl)(phenyl)methanamine 97%, Ambeed.Inc

Supplier: Ambeed
Warning

A279728-1G A279728-5G A279728-250MG
77014-592EA 116.7 USD
77014-592 77014-594 77683-844
(4-Bromophenyl)(phenyl)methanamine 97%, Ambeed.Inc
(4-Bromophenyl)(phenyl)methanamine

Precaución: For research use only.

Formula: C₁₃H₁₂BrN
MW: 262.14 g/mol
MDL Number: MFCD06616841
CAS Number: 55095-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR