SB-715992 98%

Supplier: Ambeed
Warning

Synonyms: (R)-N-(3-Aminopropyl)-N-(1-(3-benzyl-7-chloro-4-oxo-3,4-dihydroquinazolin-2-yl)-2-methylpropyl)-4-methylbenzamide, Ispinesib

A279561-1MG A279561-5MG A279561-250MG A279561-100MG A279561-1G A279561-50MG
77032-140EA 22 USD
77032-140 77032-142 77571-740 77668-234 77872-343 77872-344
SB-715992 98%
SB-715992

Precaución: for Research Use Only

Formula: C₃₀H₃₃ClN₄O₂
MDL Number: MFCD22628831
CAS Number: 336113-53-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR