5-Bromo-2-(tert-butoxy)pyridine 95%

Supplier: Ambeed
Warning

A279550-1G A279550-250MG A279550-5G
77014-586EA 130.5 USD
77014-586 77014-588 77014-590
5-Bromo-2-(tert-butoxy)pyridine 95%
5-Bromo-2-(tert-butoxy)pyridine

Precaución: for Research Use Only

Formula: C₉H₁₂BrNO
MW: 230.10 g/mol
MDL Number: MFCD08689444
CAS Number: 850495-91-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR