3-Fluoro-5-(trifluoromethyl)pyridine-2-carboxylic acid 97%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: 3-Fluoro-5-(trifluoromethyl)picolinic acid

A279376-250MG A279376-1G
76988-458EA 39 USD
76988-458 77683-843
3-Fluoro-5-(trifluoromethyl)pyridine-2-carboxylic acid 97%, Ambeed.Inc
3-Fluoro-5-(trifluoromethyl)pyridine-2-carboxylic acid

Precaución: For research use only.

Formula: C₇H₃F₄NO₂
MW: 209.1 g/mol
MDL Number: MFCD11036125
CAS Number: 89402-28-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR