Methyl 4-amino-2-methylthiophene-3-carboxylate 97%

Supplier: Ambeed
Warning

A279232-100MG A279232-1G
77014-570EA 94.43 USD
77014-570 77014-572
Methyl 4-amino-2-methylthiophene-3-carboxylate 97%
Methyl 4-amino-2-methylthiophene-3-carboxylate

Precaución: for Research Use Only

Formula: C₇H₉NO₂S
MW: 171.22 g/mol
MDL Number: MFCD01570124
CAS Number: 114943-05-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR