Methyl 4-fluoropicolinate 98%

Supplier: Ambeed
Warning

A279180-100MG A279180-1G A279180-250MG A279180-5G
76988-452EA 19 USD
76988-452 76988-454 76988-456 77890-506
Methyl 4-fluoropicolinate 98%

Precaución: for Research Use Only

Formula: C₇H₆FNO₂
MDL Number: MFCD07374898
CAS Number: 886371-79-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR