3-Bromo-4-(tert-butyl)aniline 95%

Supplier: Ambeed
Warning

A279001-1G A279001-250MG
76988-450EA 181.42 USD
76988-450 77683-842
3-Bromo-4-(tert-butyl)aniline 95%
3-Bromo-4-(tert-butyl)aniline

Precaución: For research use only.

Formula: C₁₀H₁₄BrN
MW: 228.13 g/mol
MDL Number: MFCD09966047
CAS Number: 103275-21-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR