1-Benzyl-1,2,3,6-tetrahydropyridine 95%

Supplier: Ambeed
Warning

A278850-10G A278850-1G A278850-5G
77032-124EA 76.39 USD
77032-124 77032-126 77032-128
1-Benzyl-1,2,3,6-tetrahydropyridine 95%
1-Benzyl-1,2,3,6-tetrahydropyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₅N
MDL Number: MFCD11501660
CAS Number: 40240-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR