Benzyl (3-hydroxypropyl)(methyl)carbamate 95+%

Supplier: Ambeed
Warning

A278745-1G A278745-250MG A278745-100MG
77014-556EA 174 USD
77014-556 77014-558 77892-554
Benzyl (3-hydroxypropyl)(methyl)carbamate 95+%
Benzyl (3-hydroxypropyl)(methyl)carbamate

Precaución: for Research Use Only

Formula: C₁₂H₁₇NO₃
MW: 223.26 g/mol
MDL Number: MFCD09953268
CAS Number: 887757-51-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR