2-Amino-3-thiophenecarbonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 2-Aminothiophene-3-carbonitrile

A278561-10G A278561-1G A278561-5G
77014-550EA 24.41 USD
77014-550 77014-552 77014-554
2-Amino-3-thiophenecarbonitrile 98%
2-Amino-3-thiophenecarbonitrile

Precaución: for Research Use Only

Formula: C₅H₄N₂S
MW: 124.16 g/mol
MDL Number: MFCD00706298
CAS Number: 4651-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR