(S)-1-(2-Fluorophenyl)propan-1-amine hydrochloride 98%

Supplier: Ambeed
Warning

A278354-1G A278354-250MG
76988-442EA 170.81 USD
76988-442 76988-444
(S)-1-(2-Fluorophenyl)propan-1-amine hydrochloride 98%
(S)-1-(2-Fluorophenyl)propan-1-amine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₃ClFN
MW: 189.66 g/mol
MDL Number: MFCD12757044
CAS Number: 1310923-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR