6-Bromopyrazolo[1,5-a]pyrimidine-3-carbonitrile 98%

Supplier: Ambeed
Danger

A277611-1G A277611-250MG A277611-5G
77014-530EA 35 USD
77014-530 77014-532 77014-534
6-Bromopyrazolo[1,5-a]pyrimidine-3-carbonitrile 98%
6-Bromopyrazolo[1,5-a]pyrimidine-3-carbonitrile

Precaución: for Research Use Only

Formula: C₆H₃BrN₄
MDL Number: MFCD00461392
CAS Number: 352637-44-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR