Ethyl 3-oxohept-6-enoate 95%

Supplier: Ambeed
Danger

A277580-1G A277580-250MG A277580-5G
76988-432EA 89.12 USD
76988-432 76988-434 76988-436
Ethyl 3-oxohept-6-enoate 95%
Ethyl 3-oxohept-6-enoate

Precaución: for Research Use Only

Formula: C₉H₁₄O₃
MW: 170.20 g/mol
MDL Number: MFCD00026910
CAS Number: 17605-06-0
UN: 3082
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR