2-Chloro-4-(difluoromethyl)pyridine 97%

Supplier: Ambeed
Warning

A277545-100MG A277545-250MG
77014-528EA 26 USD
77014-528 77612-608
2-Chloro-4-(difluoromethyl)pyridine 97%
2-Chloro-4-(difluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₄ClF₂N
MDL Number: MFCD14525492
CAS Number: 1204296-03-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR