Fmoc-2-bromo-D-phenylalanine 98%

Supplier: Ambeed
Warning

Synonyms: (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(2-bromophenyl)propanoic acid, Fmoc-D-Phe(2-Br)-OH

A277534-1G A277534-250MG A277534-5G
77032-102EA 27.59 USD
77032-102 77032-104 77032-106
Fmoc-2-bromo-D-phenylalanine 98%
Fmoc-2-bromo-D-phenylalanine

Precaución: for Research Use Only

Formula: C₂₄H₂₀BrNO₄
MW: 466.34 g/mol
MDL Number: MFCD01311770
CAS Number: 220497-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR