2,5-Dichlorophenylacetic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-(2,5-Dichlorophenyl)acetic acid
A277457-100MG
A277457-10G
A277457-1G
A277457-250MG
A277457-25G
A277457-5G
77032-090EA
5.31
USD
77032-090
77032-092
77032-094
77032-096
77032-098
77032-100
2,5-Dichlorophenylacetic acid 98%
2,5-Dichlorophenylacetic acid
Precaución: for Research Use Only
Formula:
C₈H₆Cl₂O₂ MW: 205.04 g/mol |
MDL Number:
MFCD02664807 CAS Number: 5398-79-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...