Ethyl 5-amino-6-methylpyridine-3-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 5-amino-6-methylnicotinate

A277178-1G A277178-250MG A277178-5G
76988-422EA 47.75 USD
76988-422 76988-424 77593-706
Ethyl 5-amino-6-methylpyridine-3-carboxylate 95%
Ethyl 5-amino-6-methylpyridine-3-carboxylate

Precaución: for Research Use Only

Formula: C₉H₁₂N₂O₂
MW: 180.204 g/mol
MDL Number: MFCD18256589
CAS Number: 1008138-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR