Dibenzo[b,d]furan-4-amine 95%

Supplier: Ambeed
Warning

Synonyms: 4-Dibenzofuranamine, 4-Aminodibenzofuran

A277146-100MG A277146-10G A277146-1G A277146-250MG A277146-5G
77032-076EA 7 USD
77032-076 77032-078 77032-080 77032-082 77668-210
Dibenzo[b,d]furan-4-amine 95%
Dibenzo[b,d]furan-4-amine

Precaución: for Research Use Only

Formula: C₁₂H₉NO
MDL Number: MFCD11110988
CAS Number: 50548-43-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR