4-(1H-Pyrazol-1-ylmethyl)aniline 95%

Supplier: Ambeed
Warning

Synonyms: 4-((1H-Pyrazol-1-yl)methyl)aniline

A276845-100MG A276845-250MG A276845-5G
77002-080EA 31.83 USD
77002-080 77002-082 77002-084
4-(1H-Pyrazol-1-ylmethyl)aniline 95%
4-(1H-Pyrazol-1-ylmethyl)aniline

Precaución: for Research Use Only

Formula: C₁₀H₁₁N₃
MW: 173.22 g/mol
MDL Number: MFCD03422514
CAS Number: 142335-61-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR