Piperazine-2-carbonitrile dihydrochloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Cyanopiperazine dihydrochloride
A276737-100MG
A276737-1G
A276737-250MG
A276737-5G
77032-062EA
21
USD
77032-062
77032-064
77032-066
77032-068
Piperazine-2-carbonitrile dihydrochloride 97%
Piperazine-2-carbonitrile dihydrochloride
Precaución: for Research Use Only
Formula:
C5H11Cl2N3 MW: 184.0700 g/mol |
MDL Number:
MFCD12910518 CAS Number: 187589-35-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...