2-Bromo-5-methoxypyrimidine 98%

Supplier: Ambeed
Warning

A276645-100MG A276645-1G A276645-250MG A276645-5G A276645-25G
76988-404EA 6 USD
76988-404 76988-406 76988-408 76988-410 77885-737
2-Bromo-5-methoxypyrimidine 98%
2-Bromo-5-methoxypyrimidine

Precaución: for Research Use Only

Formula: C₅H₅BrN₂O
MDL Number: MFCD12025462
CAS Number: 1209459-99-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR