2-Chloro-5-methylisonicotinic acid 95%

Supplier: Ambeed
Warning

A276618-100MG A276618-1G A276618-250MG A276618-5G
77014-518EA 48.81 USD
77014-518 77014-520 77014-522 77014-524
2-Chloro-5-methylisonicotinic acid 95%
2-Chloro-5-methylisonicotinic acid

Precaución: for Research Use Only

Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
MDL Number: MFCD13188984
CAS Number: 951030-56-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR