1-(4-Bromophenyl)-2,2,2-trifluoroethanol 98%

Supplier: Ambeed
Warning

A276269-1G A276269-250MG
76988-400EA 175.05 USD
76988-400 76988-402
1-(4-Bromophenyl)-2,2,2-trifluoroethanol 98%
1-(4-Bromophenyl)-2,2,2-trifluoroethanol

Precaución: for Research Use Only

Formula: C₈H₆BrF₃O
MW: 255.032 g/mol
MDL Number: MFCD12786801
CAS Number: 76911-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR