2-(1H-1,2,4-Triazol-3-yl)pyridine 97%

Supplier: Ambeed
Warning

Synonyms: 3-(2-Pyridyl)-1,2,4-triazole

A276219-10G A276219-1G A276219-250MG A276219-5G
77002-066EA 196.27 USD
77002-066 77002-068 77002-070 77002-072
2-(1H-1,2,4-Triazol-3-yl)pyridine 97%
2-(1H-1,2,4-Triazol-3-yl)pyridine

Precaución: for Research Use Only

Formula: C₇H₆N₄
MW: 146.15 g/mol
MDL Number: MFCD13188589
CAS Number: 23195-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR