Methyl-1-methyl-2-oxo-2,3-dihydro-1H-1,3-benzimidazole-5-carboxylate 95%
Supplier: Ambeed
|
Warning
|
A276047-100MG
A276047-250MG
77002-062EA
40
USD
77002-062
77002-064
Methyl-1-methyl-2-oxo-2,3-dihydro-1H-1,3-benzimidazole-5-carboxylate 95%
Methyl-1-methyl-2-oxo-2,3-dihydro-1H-1,3-benzimidazole-5-carboxylate
Precaución: for Research Use Only
Formula:
C₁₀H₁₀N₂O₃ |
MDL Number:
MFCD12922711 CAS Number: 396652-38-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...