2,5-Dibromopyridin-3-ol 97%

Supplier: Ambeed
Warning

A275854-100MG A275854-1G A275854-250MG A275854-5G
77002-052EA 68.99 USD
77002-052 77002-054 77002-056 77002-058
2,5-Dibromopyridin-3-ol 97%
2,5-Dibromopyridin-3-ol

Precaución: for Research Use Only

Formula: C₅H₃Br₂NO
MDL Number: MFCD09959722
CAS Number: 857429-79-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR