6-Chloro-3,4-dihydro-2H-isoquinolin-1-one 98%

Supplier: Ambeed
Warning

A275850-5G A275850-1G A275850-250MG
77014-516EA 234 USD
77014-516 77892-537 77892-538
6-Chloro-3,4-dihydro-2H-isoquinolin-1-one 98%
6-Chloro-3,4-dihydro-2H-isoquinolin-1-one

Precaución: for Research Use Only

Formula: C₉H₈ClNO
MDL Number: MFCD04114866
CAS Number: 22246-02-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR