N-Boc-Piperidin-4-ylacetic acid methyl ester 95%

Supplier: Ambeed
Warning

Synonyms: N-Boc-Piperidin-4-yl-acetic acid methyl ester, Methyl (piperidin-4-yl)acetate, N-BOC protected

A275715-100G A275715-10G A275715-1G A275715-25G A275715-5G
77002-044EA 111 USD
77002-044 77002-046 77002-048 77002-050 77672-416
N-Boc-Piperidin-4-ylacetic acid methyl ester 95%
N-Boc-Piperidin-4-ylacetic acid methyl ester

Precaución: for Research Use Only

Formula: C₁₃H₂₃NO₄
MDL Number: MFCD01632529
CAS Number: 175213-46-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR