5,7-Difluorochroman-4-amine 97%

Supplier: Ambeed
Danger

A275626-1G A275626-250MG A275626-5G
77032-030EA 203.7 USD
77032-030 77032-032 77032-034
5,7-Difluorochroman-4-amine 97%
5,7-Difluorochroman-4-amine

Precaución: for Research Use Only

Formula: C₉H₉F₂NO
MW: 185.17 g/mol
MDL Number: MFCD07374048
CAS Number: 886762-87-4
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR