Boc-D-Phe(4-NHfmoc)-OH 97%

Supplier: Ambeed
Warning

Synonyms: Boc-p-amino-D-Phe(Fmoc)-OH, (R)-3-(4-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)phenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid

A275618-1G A275618-5G
77002-042EA 19 USD
77002-042 77881-751
Boc-D-Phe(4-NHfmoc)-OH 97%
Boc-D-Phe(4-NHfmoc)-OH

Precaución: for Research Use Only

Formula: C₂₉H₃₀N₂O₆
MW: 502.567 g/mol
MDL Number: MFCD00151888
CAS Number: 173054-11-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR