6-Chloro-3-methylpicolinic acid 95%

Supplier: Ambeed
Warning

A275183-250MG A275183-25G A275183-5G A275183-1G
77032-014EA 22.28 USD
77032-014 77032-016 77032-018 77601-218
6-Chloro-3-methylpicolinic acid 95%
6-Chloro-3-methylpicolinic acid

Precaución: for Research Use Only

Formula: C₇H₆ClNO₂
MDL Number: MFCD16249840
CAS Number: 1201924-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR