4-Bromo-3-chloro-2-fluoropyridine 98%

Supplier: Ambeed
Warning

A275083-100MG A275083-1G A275083-250MG A275083-5G
76988-362EA 14.86 USD
76988-362 76988-364 76988-366 76988-368
4-Bromo-3-chloro-2-fluoropyridine 98%
4-Bromo-3-chloro-2-fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₂BrClFN
MDL Number: MFCD11040607
CAS Number: 1017793-21-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR