N-(2-Aminoethyl)maleimide, trifluoroacetate salt 95%

Supplier: Ambeed
Warning

Synonyms: 1-(2-Aminoethyl)-1H-pyrrole-2,5-dione 2,2,2-trifluoroacetate, Mal-NH2 TFA

A275035-100MG A275035-1G A275035-250MG A275035-5G
77032-008EA 12 USD
77032-008 77032-010 77032-012 77672-414
N-(2-Aminoethyl)maleimide, trifluoroacetate salt 95%
N-(2-Aminoethyl)maleimide, trifluoroacetate salt

Precaución: for Research Use Only

Formula: C₆H₉N₂O₂·CF₃CO₂
MW: 254.163 g/mol
MDL Number: MFCD01073617
CAS Number: 146474-00-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR