Methyl 5-bromo-2,3-dihydroxybenzoate 98%

Supplier: Ambeed
Warning

A274600-100MG A274600-250MG
76988-356EA 30.77 USD
76988-356 76988-358
Methyl 5-bromo-2,3-dihydroxybenzoate 98%
Methyl 5-bromo-2,3-dihydroxybenzoate

Precaución: for Research Use Only

Formula: C₈H₇BrO₄
MW: 247.04 g/mol
MDL Number: MFCD01412199
CAS Number: 105603-49-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR