5-Methoxybenzoxazole-2-thiol 95%

Supplier: Ambeed
Danger

A274587-10G A274587-1G A274587-250MG A274587-5G A274587-100MG
77031-996EA 644 USD
77031-996 77031-998 77032-000 77032-002 77872-331
5-Methoxybenzoxazole-2-thiol 95%
5-Methoxybenzoxazole-2-thiol

Precaución: for Research Use Only

Formula: C₈H₇NO₂S
MDL Number: MFCD03030041
CAS Number: 49559-83-3
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR