4-(Aminomethyl)-1H-indazole 98%

Supplier: Ambeed
Warning

Synonyms: (1H-Indazol-4-yl)methanamine, (1H-Indazol-4-yl)methylamine

A274575-1G A274575-250MG
77031-994EA 258.86 USD
77031-994 77571-686
4-(Aminomethyl)-1H-indazole 98%
4-(Aminomethyl)-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₉N₃
MW: 147.177 g/mol
MDL Number: MFCD10696781
CAS Number: 944898-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR