5,6-Dichloro-1H-indazole-3-carboxylic acid 96%

Supplier: Ambeed
Warning

A274516-100MG A274516-1G A274516-250MG
77014-492EA 108.22 USD
77014-492 77014-494 77014-496
5,6-Dichloro-1H-indazole-3-carboxylic acid 96%
5,6-Dichloro-1H-indazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₄Cl₂N₂O₂
MW: 231.04 g/mol
MDL Number: MFCD05663988
CAS Number: 124459-91-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR