(2S,3R,4S,5S,6R)-2-(2-Chloro-4-nitrophenoxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol 95%
Supplier: Ambeed
|
Warning
|
A274483-100MG
A274483-250MG
76988-346EA
44.56
USD
76988-346
76988-348
(2S,3R,4S,5S,6R)-2-(2-Chloro-4-nitrophenoxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol 95%
(2S,3R,4S,5S,6R)-2-(2-Chloro-4-nitrophenoxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol
Precaución: for Research Use Only
Formula:
C₁₂H₁₄ClNO₈ MW: 335.69 g/mol |
MDL Number:
MFCD00037475 CAS Number: 120221-14-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...