2,2'-(2,5-Dimethyl-1,4-phenylene)bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane) 98%

Supplier: Ambeed
Warning

A274344-1G A274344-250MG A274344-5G
77014-486EA 201.58 USD
77014-486 77014-488 77014-490
2,2'-(2,5-Dimethyl-1,4-phenylene)bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane) 98%
2,2'-(2,5-Dimethyl-1,4-phenylene)bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane)

Precaución: for Research Use Only

Formula: C₂₀H₃₂B₂O₄
MW: 358.08 g/mol
MDL Number: MFCD11855970
CAS Number: 303006-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR