Fmoc-N-Me-D-Leu-OH 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-Fmoc-N-methyl-D-leucine, (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-4-methylpentanoic acid, Fmoc-D-N-Me-Leu-OH
A273782-10G
A273782-1G
A273782-250MG
A273782-5G
77031-986EA
166.57
USD
77031-986
77031-988
77031-990
77031-992
Fmoc-N-Me-D-Leu-OH 97%
Fmoc-N-Me-D-Leu-OH
Precaución: for Research Use Only
Formula:
C₂₂H₂₅NO₄ MW: 367.445 g/mol |
MDL Number:
MFCD00235877 CAS Number: 103478-63-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...