5-(Trifluoromethyl)pyridine-2-carboxylic acid methyl ester 95%

Supplier: Ambeed
Warning

A273590-10G A273590-1G A273590-250MG A273590-25G A273590-5G
77014-476EA 25 USD
77014-476 77014-478 77014-480 77014-482 77014-484
5-(Trifluoromethyl)pyridine-2-carboxylic acid methyl ester 95%
5-(Trifluoromethyl)pyridine-2-carboxylic acid methyl ester

Precaución: for Research Use Only

Formula: C₈H₆F₃NO₂
MDL Number: MFCD08741523
CAS Number: 124236-37-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR